Noshok Pressure Transmitters with Welded Pressure Chamber
Feature an advanced diffused semi-conductor and sputtered thin film sensor for maximum stability
  • Accuracy of ±0.5% full-scale
  • Welded stainless steel pressure chamber
  • Compact size and affordable price
These highly repeatable, shock-resistant transmitters offer long-term stability and accuracy. Suppressed RFI, EMI and ESD, combined with reverse polarity and over-voltage protection, ensure reliable performance for the most demanding applications. Final calibration tests prior to shipment ensures 100% "out of the box" reliability.
Other ranges and absolute pressure measurement are available. Contact an Application Specialist for details.
132,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
NOSHOK Pressure Transmitter, 1/4" NPT(M), 5 psi 6807560 Mfr # PT30-5PSIG-1-1-2-7
COMPARE
±0.5% full-scale 5
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 60 psi 6807562 Mfr # PT30-60PSIG-1-1-2-7
COMPARE
±0.5% full-scale 60
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 100 psi 6807564 Mfr # PT30-100PSIG-1-1-2-7
COMPARE
±0.5% full-scale 100
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 200 psi 6807568 Mfr # PT30-200PSIG-1-1-2-7
COMPARE
±0.5% full-scale 200
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 300 psi 6807570 Mfr # PT30-300PSIG-1-1-2-7
COMPARE
±0.5% full-scale 300
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 500 psi 6807572 Mfr # PT30-500PSIG-1-1-2-7
COMPARE
±0.5% full-scale 500
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 750 psi 6807574 Mfr # PT30-750PSIG-1-1-2-7
COMPARE
±0.5% full-scale 750
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 1000 psi 6807576 Mfr # PT30-1000PSIG-1-1-2-7
COMPARE
±0.5% full-scale 1000
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 3000 psi 6807580 Mfr # PT30-3000PSIG-1-1-2-7
COMPARE
±0.5% full-scale 3000
₹132,000.00INR / Each
NOSHOK Pressure Transmitter, 1/4" NPT(M), 5000 psi 6807582 Mfr # PT30-5000PSIG-1-1-2-7
COMPARE
±0.5% full-scale 5000
₹132,000.00INR / Each
Showing 1- 10 of 10