North Sciences VRE Series High-Performance Lab/Pharmacy Refrigerator
VRE series high-performance pharmacy refrigerators with standard drawers are designed with features that support sample protection for storage requiring 2° to 8°C, according to CDC recommendations
  • Cloud-based monitoring provided by TraceableLive® to securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Strong casters with brakes makes moving the refrigerators even easier
  • Triple coated tempered glass with heat reflective film ensures the inner temperature remains stable and is anti-condensation
  • Refrigeration block is fully replaceable
  • Forced-air circulation with special air ducts guarantees inner temperature consistency
  • Unique evaporator design results in better uniformity with no ice build-up
VRE series high-performance pharmacy refrigerators are designed with features that support sample protection for the storage of pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C, according to CDC recommendations. The controller allows the inner temperature to be adjusted by increments of 0.1° with multiple sensors to display and accurately control the temperature. A safety door lock prevents unauthorized access. Auto defrost function offers reliable performance in high ambient temperature and high humidity areas.
The refrigerators are paired with a Traceable® data logger, including a one-year subscription to TraceableLIVE® cloud-based control and monitoring services.
1,147,000.00 - ₹2,766,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature - Refrigerator (° C) Power (VAC) Refrigerator Capacity (Liters)
Availability Pricing
North Sciences VRE Series High-Performance Lab/Pharmacy Refrigerator, 2°C to 10°C, 600 L, with TraceableLive®, 110V WW-44261-63 Mfr # VRE-0500-NA
COMPARE
2 110 600
₹1,241,000.00INR / Each
North Sciences VRE Series High-Performance Lab/Pharmacy Refrigerator, 2°C to 10°C, 600 L, with TraceableLive®, 220V WW-44261-64 Mfr # VRE-0500-EU
COMPARE
2 220 600
₹1,147,000.00INR / Each
North Sciences VRE Series High-Performance Lab/Vaccine Refrigerator, 2-Door, 2°C to 10°C, 1100 L, with TraceableLive®, 110V WW-44261-67 Mfr # VRE-1000-NA
COMPARE
2 120 1100
₹2,766,000.00INR / Each
North Sciences VRE Series High-Performance Lab/Vaccine Refrigerator, 2-Door, 2°C to 10°C, 1100 L, with TraceableLive®, 220V WW-44261-68 Mfr # VRE-1000-EU
COMPARE
2 220 1100
₹2,449,000.00INR / Each