Nichrome V Calibrated Inoculating Loops
Select, inoculate, or transfer microbiological samples
  • Nichrome wire composition cools quickly
  • Constructed of 26-gauge wire
Ideal to streak various liquids, suspensions, bacteria, and other specimens on to culture media and/or petri dishes for purposes of study and growth. Each loop is calibrated to APHA standards for optimal accuracy and reproducibility. Choose from loops with or without handle. Order handle 14203-34 separately for loops without handle included.
4,800.00 - ₹8,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Contained Volume (µL) Handle Type Loop Length (in)
Availability Pricing
Nichrome V Inoculating Loop with Insulated Handle, Calibrated, 26 Gauge, 1 µL 1420325 Mfr # 3730
COMPARE
1 Insulated 1 3/4
₹8,300.00INR / Each
Nichrome V Inoculating Loop with Insulated Handle, Calibrated, 26 Gauge, 10 µL 1420326 Mfr # 3740
COMPARE
10 Insulated 1 3/4
₹4,800.00INR / Each
Nichrome V Inoculating Loop without Handle, Calibrated, 26 Gauge, 1 µL 1420331 Mfr # 3630
COMPARE
1 No Handle 3
₹5,100.00INR / Each