National Optical Digital Compound Microscopes with Tablet

Capture, view, edit, and share live images with Wi-Fi enabled camera and Android™ tablet
  • Documentation and analysis for a deeper view of your slide
  • Simply log on with your tablet and begin to capture, edit, and save your images
  • Perfect for the immediate presentation of image results
  • 8" or 10” LCD Android™ tablet with 1280 x 800 screen resolution
National Optical digital compound microscopes with tablet is designed to display images and video directly on the tablet or transfer to Wi-Fi enabled devices. This setup is perfect for the immediate presentation of image results to colleagues. Detaching the tablet for an even more flexible application is possible thanks to the Wi-Fi data transfer. Use the pre-installed MotiConnect App and get ready to capture, edit and share your microscope images from the tablet in a few taps. These cameras can produce still image and video capture, live and still image measurement, and image adjustments.

Choose from microscopes with an 8” or 10” tablet, corded or cordless power, 400X or 1000X magnification, and different objective types.
231,000.00 - ₹603,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Objectives Size
Availability Pricing
National Optical Digital Compound Microscope with 8" Tablet, Monocular, 4X/10X/40X, Coaxial Focusing Controls; Cordless 4892634 Mfr # BTI1-213-LED
COMPARE
Monocular 4X, 10X, 40XR 8" 1280 x 800 Tablet
₹280,000.00INR / Each
National Optical Digital Compound Microscope with 8" Tablet, Monocular, 4X/10X/40X, Separate Focusing Controls; Cordless 4892635 Mfr # BTI1-205-LED
COMPARE
Monocular 4X, 10X, 40X 8" 1280 x 800 Tablet
₹231,000.00INR / Each
National Optical Digital Compound Microscope with 8" Tablet, Trinocular, 4X/10X/40X/100X, Super High Contrast Lens; 110-240 VAC 4892636 Mfr # BTI1-169-ASC
COMPARE
Trinocular 4X, 10X, 40X , 100X Super High Contrast 8" 1280 x 800 Tablet
₹522,000.00INR / Each
National Optical Digital Compound Microscope with 8" Tablet, Trinocular, 4X/10X/40X/100X, Plan Lens; 110-240 VAC 4892637 Mfr # BTI1-169-P
COMPARE
Trinocular 4X, 10X, 40X , 100X Plan 8" 1280 x 800 Tablet
₹561,000.00INR / Each
National Optical Digital Compound Microscope with 10" Tablet, Trinocular, 4X/10X/40X/100X, Plan Lens; 110-240 VAC 4892638 Mfr # BTI2-169-P
COMPARE
Trinocular 4X, 10X, 40X , 100X Plan 10" 1280 x 800 Tablet
₹603,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat