National Optical 440-Series Professional Stereozoom Microscopes
Robust and sturdy microscope that provides crisp, clear images
  • Large-scale 6:1 zoom ratio offers clear distortion-free images
  • Comfortable 45° viewing angle minimizes strain, even after long hours
  • Ergonomically designed with durable metal body ensures years of consistent performance without compromising quality
National Optical 440-series professional stereozoom microscopes feature a large zoom ratio of 6:1 and are ideal for industrial and educational applications. The 0.75-4.5X eyepieces combine with the 10X objective for a magnification range of 7.5-45X. Microscopes feature a heavy-duty post mount with locking support collar to permit up or down adjustment of viewing head to accommodate different sized objects on stage. The overall height is adjustable from 13.4” to 17” to accommodate larger objections.
Both binocular and trinocular models are available with or without LED illumination. Models with LED illumination feature top and bottom LED and base mounted light controls to permit choice of top only, bottom only, or dual illumination.
181,000.00 - ₹270,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Illuminator(s) Magnification
Availability Pricing
National Optical Professional Stereozoom Microscope, Binocular, Dual LED Base; 110-240 VAC 4892639 Mfr # 440-440PLL
COMPARE
Binocular Dual LED 7.5X to 45X
₹237,000.00INR / Each
National Optical Professional Stereozoom Microscope, Binocular, Plain Base; 110-240 VAC 4892640 Mfr # 440-440P
COMPARE
Binocular None 7.5X to 45X
₹181,000.00INR / Each
National Optical Professional Stereozoom Microscope, Trinocular, Dual LED Base; 110-240 VAC 4892641 Mfr # 440T-440PLL
COMPARE
Trinocular Dual LED 7.5X to 45X
₹270,000.00INR / Each
National Optical Professional Stereozoom Microscope, Trinocular, Plain Base; 110-240 VAC 4892642 Mfr # 440T-440P
COMPARE
Trinocular None 7.5X to 45X
₹219,000.00INR / Each