National Optical 214-RLED Compound Monocular Microscope
Cordless microscope uses rechargeable batteries for added versatility
  • Energy efficient, rechargeable LED illumination allows up to 50 hours of use on a single 8 hour charge
  • Camera port integrated into head for attachment of optional Moticam camera
  • 45° inclined monocular viewing head
  • Indicator light changes from red to green when fully charged
The National Optical 214-RLED compound monocular microscope features a versatile design that includes cordless battery operation, built-in camera port, and 4-hole objective turret (nosepiece). The objective turret is in reverse position to permit easier changing and positioning of specimen slides. The rechargeable LED illumination system provides optimum image brightness, virtually no bulb heat, and has a 50,000-hour rated bulb life. Batteries allow up to 500 recharges, for a total of 20,000 to 25,000 hours use from a single set of 3 AA rechargeable nickel hydride batteries. Integrated camera port allows for easy installation of a Moticam camera for image capture. C-mount adapter and camera sold separately.
159,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Illuminator(s) Magnification Power
Availability Pricing
National Optical Compound Monocular Microscope with 4-Hole Objective Turret and Camera Port, 4X/10X/40X/100X 4892623 Mfr # 214-RLED
COMPARE
LED 40X/100X/400X/1000X AA rechargeable nickel hydride batteries
₹159,000.00INR / Each