National Optical 167-Series LED Compound Microscopes
Superb optical clarity and reliable mechanical engineering assure years of top performance
  • Variable LED illumination provides optimal image brightness
  • Head rotates 360° for ergonomic viewing and easy eyepiece sharing
  • Rugged cast aluminum frame provides enhanced durability
The National Optical 167-series compound microscopes incorporate a wide range of features designed for university and laboratory use. Superb optical clarity and reliable mechanical engineering assure years of top performance. The quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives and features textured rubber knurling for easy grip. The objective turret is in reverse position to permit easier changing and positioning of specimen slides. 360° swiveling eyepiece offers a comfortable observation position, especially for prolonged periods.
162,000.00 - ₹319,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Objectives
Availability Pricing
National Optical Compund Microscope, LED, Trinocular, 4X/10X/40X/100X, Semi-Plan Lens; 110-220 VAC 4892627 Mfr # 169-SP
COMPARE
Trinocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Semi-Plan
₹272,000.00INR / Each
National Optical Compund Microscope, LED, Trinocular, 4X/10X/40X/100X, Super High Contrast Lens; 110-220 VAC 4892628 Mfr # 169-ASC
COMPARE
Trinocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Super High Contrast
₹286,000.00INR / Each
National Optical Compund Microscope, LED, Trinocular, 4X/10X/40X/100X, Plan Lens; 110-220 VAC 4892629 Mfr # 169-P
COMPARE
Trinocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Plan
₹319,000.00INR / Each
National Optical Compund Microscope, LED, Binocular, 4X/10X/40X/100X, Super High Contrast Lens; 110-220 VAC 4892630 Mfr # 168-ASC
COMPARE
Binocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Super High Contrast
₹224,000.00INR / Each
National Optical Compund Microscope, LED, Binocular, 4X/10X/40X/100X, Plan Lens; 110-220 VAC 4892631 Mfr # 168-P
COMPARE
Binocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Plan
₹257,000.00INR / Each
National Optical Compund Microscope, LED, Monocular, 4X/10X/40X/100X, Super High Contrast Lens; 110-220 VAC 4892632 Mfr # 167-ASC
COMPARE
Monocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Super High Contrast
₹175,000.00INR / Each
National Optical Compund Microscope, LED, Monocular, 4X/10X/40X/100X, Semi-Plan Lens; 110-220 VAC 4892633 Mfr # 167-SP
COMPARE
Monocular 40X/100X/400X/1000X 4X, 10X, 40X , 100X Semi-Plan
₹162,000.00INR / Each
Showing 1- 7 of 7