Nabertherm Basic Muffle Furnace
Compact, lightweight design is ideal for crowded labs
  • Reach temperatures up to 1100°C
  • Quartz glass tube heating elements provide uniform heating from two sides
  • Rust-free stainless steel exterior and dual shell housing for low external temperatures
This muffle furnace features a space-saving under-door controller and solid-state relays for quiet operation. Flap-style door provides easy access to furnace chamber and can double as a work platform. Hot air is expelled through exhaust outlet in the rear wall of the furnace.
1,164,000.00 - ₹2,291,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC)
Availability Pricing
Nabertherm LE2/11 Muffle Furnace with R7 Controller, 2L; 120V 3333527 Mfr # LE022K17N1-120
COMPARE
2 120
₹1,164,000.00INR / Each
Nabertherm LE2/11 Muffle Furnace with R7 Controller, 2L; 240V 3333528 Mfr # LE022K17N1-240
COMPARE
2 240
₹1,164,000.00INR / Each
Nabertherm LE6/11 Muffle Furnace with R7 Controller, 6L; 120V 3333529 Mfr # LE062K17N1-120
COMPARE
6 120
₹1,387,000.00INR / Each
Nabertherm LE6/11 Muffle Furnace with R7 Controller, 6L; 240V 3333530 Mfr # LE062K17N1-240
COMPARE
6 240
₹1,387,000.00INR / Each
Nabertherm LE14/11 Muffle Furnace with R7 Controller, 14L; 240V 3333531 Mfr # LE142K17N1-240
COMPARE
14 240
₹1,659,000.00INR / Each
Nabertherm LE24/11 Muffle Furnace with R7 Controller, 24L; 240V 3333532 Mfr # LE241K17N1-240
COMPARE
24 240
₹2,291,000.00INR / Each