Myron L Digital Conductivity/TDS Controllers
In-line stainless steel cells can withstand temperatures up to 212°F (100°C) and pressures up to 100 psi!
  • Feature a heavy-duty 10 amp relay
  • NEMA 4X (IP65) ABS enclosure
Use these conductivity/TDS controllers to monitor and control all of your conductivity processes. Order conductivity cells seperately.
182,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Temperature Compensation Max Temperature (° C) Description
Availability Pricing
Myron L 758II-109 Digital Conductivity/TDS Controller, 0 to 20.00 µS 1901016 Mfr # 758II-109
COMPARE
Automatic or Manual 100 Digital Conductivity/TDS Controller, 0 to 20.00 µS
₹182,000.00INR / Each
Myron L 758II-121 Digital Conductivity/TDS Controller, 0 to 2000 µS 1901020 Mfr # 758II-121
COMPARE
Automatic or Manual Digital Conductivity/TDS Controller, 0 to 2000 µS
₹182,000.00INR / Each