MTC Bio nUVaClean™ UV Sterilizing Pipettor Rack
Germicidal UV-C lamp decontaminates pipettors
  • UV-C lamp and reflector system destroys 99% of DNA
  • Sterilize up to six single-channel pipettors at once
  • Compact 6.5" (17 cm) diameter footprint
Destroy up to 99% of microbiological contaminants from pipettor shafts using the nUVaClean sterilizing rack. Rack features a UV-C lamp and a high-efficiency reflector/concentrator that provide 360° of 254 nm UV exposure to prevent cross contamination. Place up to six single-channel pipettors in the rack and press "start" to begin the process. Sterilizing cycle automatically shuts off after 28 minutes.
65,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Outlet Connections Power (VAC) Wavelength
Availability Pricing
MTC Bio nUVaClean™ UV Sterilizing Pipettor Rack; 120 VAC WW-14003-00 Mfr # P5590
COMPARE
US plug 120 254 nm
₹65,400.00INR / Each
MTC Bio nUVaClean™ UV Sterilizing Pipettor Rack; 230 VAC WW-14003-01 Mfr # P5590-E
COMPARE
EU, UK, or AU plug 230 254 nm
₹65,400.00INR / Each