Motic® SMZ-160 Stereozoom Microscopes
Greenough optical system allows you to move between smaller and larger samples with minimal focus adjustment
  • Large-scale 6:1 zoom ratio offers clear distortion-free images
  • Comfortable 45° viewing angle minimizes strain, even after long hours
  • Ergonomically designed with durable metal body ensures years of consistent performance without compromising quality
  • LED incident and transmitted illumination with intensity control provides optimal sample viewing
Motic® SMZ-160 stereozoom microscopes can handle the rigorous demands of industrial and educational usage while also meeting the high-performance requirements for industrial quality control, inspection, and research applications. Designed with a working distance of 100 mm and large zoom ratio of 6:1, the Greenough optical system allows you to move from smaller to larger samples and vice-versa with minimal focus adjustment. When combined with its high-eyepoint 10X/20mm eyepieces and a 45-degree inclination, this makes for hours of comfortable sample viewing.
LED illuminated stand offers a bright 3W LED incident and transmitted illumination. The provided LED light source resembles the color temperature of natural light, ensuring sample colors remain true to life. It also features intensity control for optimal viewing of all samples.
271,000.00 - ₹308,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Power (VAC)
Availability Pricing
Motic Instruments SMZ-160-BLED Stereozoom Binocular Microscope with LED Incident and Transmitted Illuminators; 100-240 VAC 4892588 Mfr # 1100201300171
COMPARE
Binocular 7.5X to 45X 100 to 240
₹271,000.00INR / Each
Motic Instruments SMZ-160-TLED Stereozoom Trinocular Microscope with LED Incident and Transmitted Illuminators; 100-240 VAC 4892589 Mfr # 1100201300181
COMPARE
Trinocular 7.5X to 45X 100 to 240
₹308,000.00INR / Each