Motic® Panthera Compound Microscopes
Ergonomically designed durable body ensures years of consistent performance without compromising quality
  • UC Plan achromatic objectives feature the color-corrected infinity optical system (CCIS®) for improved image quality
  • Rotatable, adjustable eyepiece tubes provide operator comfort
  • Adjustable interpupillary distance and ergonomic viewing angle reduce fatigue
  • Illumination with intensity control provides optimal image brightness
Motic® Panthera compound microscopes feature Ultra Contrast (UC) Plan Achromatic objectives in a range from 4X to 100X magnification. These objectives provide ultra-high contrast and very flat field of view, which together with the UC Widefield eyepieces brings you bright, detailed, and sharp images for new insights. The automated light management system features a digital intensity knob, coded LED nosepiece, and illuminator, which combine to offer information about light intensity, the microscope mode, as well as memorizing the light intensity of each objective. Other features include a reversed 4-fold nosepiece, rackless X/Y-stage with standard slide holder, and upper limit stop.
398,000.00 - ₹1,158,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Illuminator(s) Output
Availability Pricing
Motic Instruments Panthera E2 Compound Binocular Microscope, LED, 40-1000X; 100 to 240 VAC 4892582 Mfr # 1100104601116
COMPARE
Binocular 1W LED USB 2.0 for external camera power
₹398,000.00INR / Each
Motic Instruments Panthera E2 Compound Trinocular Microscope, LED, 40-1000X; 100 to 240 VAC 4892583 Mfr # 1100104601126
COMPARE
Trinocular 1W LED USB 2.0 for external camera power
₹481,000.00INR / Each
Motic Instruments Panthera C2 Compound Binocular Microscope, LED and Halogen, 40-1000X; 100 to 240 VAC 4892584 Mfr # 1100104601136
COMPARE
Binocular Interchangable 3W LED / 30W Halogen USB 2.0 for external camera power
₹551,000.00INR / Each
Motic Instruments Panthera C2 Compound Trinocular Microscope, Simple Phase Package, LED and Halogen, 40-400X; 100 to 240 VAC 4892585 Mfr # 1100104601161
COMPARE
Trinocular Interchangable 3W LED / 30W Halogen USB 2.0 for external camera power
₹711,000.00INR / Each
Motic Instruments Panthera C2 Compound Trinocular Microscope, Phase Package, LED and Halogen, 40-400X; 100 to 240 VAC 4892586 Mfr # 1100104601171
COMPARE
Trinocular Interchangable 3W LED / 30W Halogen USB 2.0 for external camera power
₹818,000.00INR / Each
Motic Instruments Panthera Cloud Compound Binocular Microscope, Smart CAM Camera, LED and Halogen, 40-1000X; 100 to 240 VAC 4892587 Mfr # 1100403300201
COMPARE
Binocular Interchangable 3W LED / 30W Halogen USB 2.0 for external camera power, HDMI, Wi-Fi, Ethernet, Cloud storage
₹1,158,000.00INR / Each