Motic® BA210S Compound Microscopes
Ergonomically designed with durable metal body ensures years of consistent performance without compromising quality
  • Plan achromatic objectives feature the color-corrected infinity optical system (CCIS®) for improved image quality
  • Equipped with a Siedentopf type head with 30° incline that rotates 360° for ergonomic viewing
  • LED illumination with intensity control provides optimal image brightness
The Motic® BA210S compound microscope is designed to provide optimal image quality in a durable, long-lasting design. The 360° swiveling eyepiece offers a comfortable observation position, especially for prolonged periods. Hard-coated stage allows a safe and durable evaluation of diverse samples. LED illumination with intensity control offers safe, long-term illumination with reduced heat production.
308,000.00 - ₹318,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Power (VAC)
Availability Pricing
Motic Instruments BA210S Biological Compound Binocular Microscope, LED, 40-1000X; 100 to 240 VAC 4892580 Mfr # 1100100403831
COMPARE
Binocular 40X, 100X, 400X, 1000X 100 to 240
₹308,000.00INR / Each
Motic Instruments BA210S Biological Compound Trinocular Microscope, LED, 40-1000X; 100 to 240 VAC 4892581 Mfr # 1100100403841
COMPARE
Trinocular 40X, 100X, 400X, 1000X 100 to 240
₹318,000.00INR / Each