Motic® AE31E LED Inverted Microscope
Designed for routine live cell inspection with an easy and flexible setup
  • Plan achromatic objectives feature the color-corrected infinity optical system (CCIS®) and Phase contrast for improved image quality
  • Equipped with a Siedentopf type head with 45° incline that rotates 360° for ergonomic viewing
  • Ideal instrument for routine live cell observation and other microbial samples
  • Easily transition from Halogen to LED illumination for optimal viewing
The Motic® AE31E inverted microscope is designed to provide superior image quality when viewing cell cultures in microbiology, living organisms, and tissue culture samples. This microscope works with the newest generation of LWD Plan Achromatic lenses which feature the color-corrected infinity optical system (CCIS®) and Phase contrast for remarkable image quality.
The 360° swiveling eyepiece offers a comfortable observation position, especially for prolonged periods. Microscopes feature an auto on/off function of the power supply, which extends the lifespan of the sample and light bulb and saves energy. The modular interchangeable illumination concept allows for the easy transition from Halogen to LED to fulfill a wide-range of viewing expectations. An anti-fungus treatment is applied to all parts of the microscope to protect the system in humid environments.
1,042,000.00 - ₹1,176,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Illuminator(s) Magnification
Availability Pricing
Motic Instruments AE31E Inverted Binocular Microscope, LED or Halogen, 40 to 400X; 100-240 VAC 4892573 Mfr # 1100100200861
COMPARE
Binocular 6V/30W Quartz Halogen or 3W LED 40X/100X/200X/400X
₹1,042,000.00INR / Each
Motic Instruments AE31E Inverted Trinocular Microscope, LED or Halogen, 40 to 400X; 100-240 VAC 4892574 Mfr # 1100100200841
COMPARE
Trinocular 6V/30W Quartz Halogen or 3W LED 40X/100X/200X/400X
₹1,176,000.00INR / Each