Motic® AE2000 Inverted Microscopes
Designed for routine live cell inspection with an easy and flexible setup
  • Plan achromatic objectives feature the color-corrected infinity optical system (CCIS®) for improved image quality
  • Equipped with a Siedentopf type head with 45° incline that rotates 360° for ergonomic viewing
  • Ideal instrument for routine live cell inspection
The Motic® AE2000 inverted microscope is designed to provide superior image quality when viewing cell cultures in microbiology, living organisms and tissue culture samples. The 360° swiveling eyepiece offers a comfortable observation position, especially for prolonged periods. Microscopes feature an auto on/off function of the power supply, which extends the lifespan of the sample and light bulb and saves energy. The modular interchangeable illumination concept, allows for the easy transition from Halogen to LED to fulfill a wide-range of viewing expectations.
715,000.00 - ₹933,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Objectives
Availability Pricing
Motic Instruments AE2000 Inverted Binocular Microscope with LED, Basic Package; 100-240 VAC 4892569 Mfr # 1100103800222
COMPARE
Binocular 40x to 100x CCIS® Plan Achromatic 4X and PH 10X
₹715,000.00INR / Each
Motic Instruments AE2000 Inverted Binocular Microscope with LED, PL PH 20X Package; 100-240 VAC 4892570 Mfr # 1100103800268
COMPARE
Binocular 40x to 200x CCIS® Plan Achromatic 4X, PH 10X, and PH 20X
₹804,000.00INR / Each
Motic Instruments AE2000 Inverted Trinocular Microscope with LED, Basic Package; 100-240 VAC 4892571 Mfr # 1100103800266
COMPARE
Trinocular 40x to 100x CCIS® Plan Achromatic 4X and PH 10X
₹844,000.00INR / Each
Motic Instruments AE2000 Inverted Trinocular Microscope with LED, PL PH 20X Package; 100-240 VAC 4892572 Mfr # 1100103800267
COMPARE
Trinocular 40x to 200x CCIS® Plan Achromatic 4X, PH 10X, and PH 20X
₹933,000.00INR / Each