MLA Digital Adjustable Volume Pipettors
Calibration feature is handy for viscous solutions
These pipettes feature special benefits without compromising MLA quality. Built with aluminum alloy and stainless steel, the lifetime warranty ensures maximum durability and performance. Pipets are engraved with serial numbers for easy traceability and quality control. Adjust or calibrate pipettors above or below the stated volume ±10%.
Ideal for laboratories performing tests requiring multiple or non-standard volumes. Easy-to-read digital volume settings with “click-stops” provide fast and easy volume selection. Low spring force minimizes thumb muscle activity.
110,000.00 - ₹117,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (mL)
Availability Pricing
VistaLab 1136 Adjustable Volume Pipette, 10-50 uL, Green; Each WW-06199-39 Mfr # 1136
COMPARE
±2.00% at 50.00µL; ±3.00% at 30.00µL; ±5.00% at 10.00µL Green 0.05
₹110,000.00INR / Each
VistaLab 1138 Adjustable Volume Pipette, 200-1000 uL, Grey; Each WW-06199-43 Mfr # 1138
COMPARE
±1.00% at 1000µL; ±1.00% at 600.0µL; ±1.00% at 200.0µL Gray 1
₹117,000.00INR / Each