Mini Needle Valves, SS
Compact and rugged—provides robust performance in the toughest applications
  • 100% helium leak-tested to 1 x 104 mL/s for guaranteed performance and reliability
  • FKM O-ring seal and PTFE back-up ring offer protection from corrosion and galling
  • Rolled stem threads provide strength and ease of operation
  • Rated for use up to 10,000 psi at 93°C (200°F)
These compact stainless steel needle valves deliver maximum durability and reliable performance in the most demanding applications. Each unit, 100% helium leak-tested to 1 x 104 mL/s, provides exceptional integrity and consistent operation. Use these hard seat valves in high-pressure applications up to 10,000 psi at 93°C (200°F). The patented metal-to-metal body-to-bonnet seal significantly expands the pressure range of the valve.
11,900.00 - ₹14,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Pattern Inlet Port Outlet Port
Availability Pricing
Needle Valve, SS, 1/4" NPT(F) x 1/4" NPT(F), Straight Flow 4852705 Mfr # 102FFS
COMPARE
Straight 1/4" NPT(F) 1/4" NPT(F)
₹12,000.00INR / Each
Needle Valve, SS, 1/4" NPT(M) x 1/4" NPT(F), 90° Elbow Flow 4852707 Mfr # 102MFAS
COMPARE
90° Elbow 1/4" NPT(M) 1/4" NPT(F)
₹14,900.00INR / Each
Needle Valve, SS, 1/4" NPT(M) x 1/4" NPT(F), Straight Flow 4852709 Mfr # 102MFS
COMPARE
Straight 1/4" NPT(M) 1/4" NPT(F)
₹11,900.00INR / Each