Microplate Orbital Shaker with Timer
Ideal for mixing test solutions in microtitration assays
  • Securely holds four 96-well plates securely with stainless steel spring clamps
Shaker applications include hematology, urology, or EIA/ELISA immunology. Chemical resistanct contoured stainless steel platform holds plates securely.
What''''s included: a platform and a 6-1/4-ft cord with U.S. standard plug
290,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Platform Length (in) Platform Width (in)
Availability Pricing
Labnet S2020-P4-220 Microplate Orbital Shaker with Timer, 230 VAC, 50 Hz 5140205 Mfr # S2020-P4-B
COMPARE
1200 8.5 7.5
₹290,000.00INR / Each