Mettler Toledo XPR Essential Analytical Balances
Offers fast, highly accurate weighing results
  • LevelControl warns when balance is not level
  • Draft shield doors open automatically with push of a button
  • Overload protection for load cell
  • User management and password protection
  • Multiple ports offer secure connectivity and data export
  • Color touch screen provides clear visibility — easy to use even with gloves on
  • StatusLight™ on terminal indicates when balance is ready for weighing
  • Fully Automatic Calibration Technology (FACT) performs internal calibrations automatically
  • SmartTrac program facilitates dosing in small increments to a target weight
Get highly accurate and fast results with the XPR Essential analytical balances. The high-performance load call and hanging weighing pan allow you to precisely weigh small samples with high accuracy, helping you reduce waste and save money. The load call and electronics are designed to withstand intensive use, providing you with years of service. The 7-inch color touch-sensitive display provides easy accessibility, can be used with gloves on, and be positioned wherever it's most convenient in the lab.
The one touch motorized doors offer effortless access to the weighing chamber making it easy to dose onto the low weighing pan. ErgoClip accessories (purchase separately) can be used to ergonomically position tare vessels. The integration with LabX software also adds a layer of automation, traceability, and data management.
The balances are also equipped with tolerance profiles to ensure weighing tasks meet defined quality requirements and regulations. In addition, the Good Weighing Practice® (GWP) Approved quality assurance function actively monitors the balance to confirm it is safe to weigh -- green light illuminates when the balance is ready to accept samples.
2,599,000.00 - ₹4,678,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Linearity (mg) Repeatability (mg)
Availability Pricing
Mettler Toledo XPR105DUE Analytical Balance, 120/41 g x 0.1/0.01 mg, Internal Calibration, SmartTrac Dosing Guide 1100381 Mfr # 30904711
COMPARE
120 0.2 0.02
₹3,564,000.00INR / Each
Mettler Toledo XPR104DUE Analytical Balance, 120/81 g x 1/0.1 mg, Internal Calibration, SmartPan Hanging Weigh Pan 1100382 Mfr # 30904736
COMPARE
120 0.2 0.1
₹2,599,000.00INR / Each
Mettler Toledo XPR225DUE Analytical Balance, 220/121 g x 0.1/0.01 mg, Internal Calibration, SmartPan Hanging Weigh Pan 1100383 Mfr # 30904667
COMPARE
220 0.2 0.02
₹4,678,000.00INR / Each
Mettler Toledo XPR205DUE Analytical Balance, 220/81 g x 0.1/0.01 mg, Internal Calibration, SmartTrac Dosing Guide 1100384 Mfr # 30904692
COMPARE
220 0.2 0.02
₹4,328,000.00INR / Each
Mettler Toledo XPR204E Analytical Balance, 220 g x 0.1 mg, Internal Calibration, SmartPan Hanging Weigh Pan 1100385 Mfr # 30904716
COMPARE
220 0.2 0.07
₹3,597,000.00INR / Each
Mettler Toledo XPR204DUE Analytical Balance, 220/121 g x 1/0.1 mg, Internal Calibration, SmartPan Hanging Weigh Pan 1100386 Mfr # 30904731
COMPARE
220 0.2 0.1
₹2,859,000.00INR / Each
Mettler Toledo XPR304E Analytical Balance, 320 g x 0.1 mg, Internal Calibration, SmartPan Hanging Weigh Pan 1100387 Mfr # 30904741
COMPARE
320 0.3 0.08
₹3,730,000.00INR / Each
Showing 1- 7 of 7