METTLER TOLEDO S2 Seven2Go™ Handheld pH/mV Meters
Get quality data with one hand
  • User-friendly interface and single-handed T-pad operation provide fast navigation
  • Rugged IP67-rated housing protects meter in harsh environments
  • Compatible with ISM sensors—no need to recalibrate with every electrode change
These handheld pH meters are perfect for fieldwork and other mobile applications. Robust construction provides durability, while the intuitive menu and simplified button configuration let you take readings quickly.
The InLab ISM (Intelligent Sensor Management) series of electrodes stores its ID number, serial number, and calibration data. When the electrode is connected to the meter it communicates this information, so there is no need to select electrode type when switching or to recalibrate every time you power on the meter.
Choose the meter alone if you already have ISM electrodes, or choose a bundled kit suitable for your application.
176,000.00 - ₹251,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo S2 Handheld pH/mV Meter Only 3590345 Mfr # 30207949
COMPARE
Handheld pH/mV Meter Only
₹176,000.00INR / Each
Mettler Toledo S2 Handheld pH/mV Meter Standard Kit 3590346 Mfr # 30207950
COMPARE
Handheld pH/mV Meter Standard Kit
₹218,000.00INR / Each
Mettler Toledo S2 Handheld pH/mV Meter Field Kit 3590347 Mfr # 30207951
COMPARE
Handheld pH/mV Meter Field Kit
₹236,000.00INR / Each
Mettler Toledo S2 Handheld pH/mV Meter Food Kit 3590348 Mfr # 30207952
COMPARE
Handheld pH/mV Meter Food Kit
₹251,000.00INR / Each