Mettler Toledo MA Series NTEP-Approved Standard Analytical Balances
Simple and efficient balances for routine everyday weighing
  • NTEP-approved legal-for-trade balances meet government standards and requirements for commercial applications
  • Large, easy-to-read LCD hybrid touchscreen simplify operation
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Internal calibration ensure accuracy
  • Flexible integration options—connect using USB-A and RS-232
  • Automatically recognizes plugged in peripheral devices
  • Easily identify samples using the sample ID function
  • Overload protection guards the weighing cell against excess weight overload
  • Remove, clean and replace all of the draft shield panels in just a few simple steps, without tools
  • High-quality materials and solid construction provide long service life
The MA series NTEP-approved analytical balances offer excellent repeatability, readability, and durability, making them perfect for routine tasks. The large, easy-to-read LCD hybrid touchscreen provides on-screen instructions for quick and efficient operation even when wearing gloves. An electromagnetic force compensation (EMFC) weighing cell and internal calibration provide consistent reliable results.
The USB-A and RS-232 interfaces allow easy transfer of data to a printer, computer, or USB storage device for complete and error-free documentation. Nine built-in applications include essential weighing functions, statistics, and formulation. Perform weighing operations below the balance with the included weighing hook. Password protection feature restricts modification of the balance settings to authorized personnel only.
Rugged housing, made of high-quality and durable materials, protects the sensitive sensor from environmental influences and impacts. Smooth surfaces and rounded edges provide quick and easy cleaning. The QuickLock system keeps draft shield panels securely in place. The draft shield is easily removed for cleaning; draft shield panels and drip tray are dishwasher safe. Built-in overload protection prolongs the life of your balance.
908,000.00 - ₹1,926,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo MA55/A Analytical Balance, NTEP-Approved, MA Series, 52g x 0.01mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262935 Mfr # 30666317
COMPARE
52 0.01 Analytical Balance, NTEP-Approved, 52 g x 0.01 mg
₹1,815,000.00INR / Each
Mettler Toledo MA155DU/A Analytical Balance, NTEP-Approved, MA Series, 62/152g x 0.01/0.1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262936 Mfr # 30697383
COMPARE
152 0.1 Dual-Range Analytical Balance, NTEP-Approved, 62/152 g x 0.01/0.1 mg
₹1,872,000.00INR / Each
Mettler Toledo MA95/A Analytical Balance, NTEP-Approved, MA Series, 92g x 0.01mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262937 Mfr # 30706702
COMPARE
92 0.01 Analytical Balance, NTEP-Approved, 92 g x 0.01 mg
₹1,926,000.00INR / Each
Mettler Toledo MA54/A Analytical Balance, NTEP-Approved, MA Series, 52g x 0.1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262938 Mfr # 30697386
COMPARE
52 0.1 Analytical Balance, NTEP-Approved, 52 g x 0.1 mg
₹908,000.00INR / Each
Mettler Toledo MA104/A Analytical Balance, NTEP-Approved, MA Series, 120g x 0.1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262939 Mfr # 30697409
COMPARE
120 0.1 Analytical Balance, NTEP-Approved, 120 g x 0.1 mg
₹977,000.00INR / Each
Mettler Toledo MA204/A Analytical Balance, NTEP-Approved, MA Series, 220g x 0.1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262940 Mfr # 30697412
COMPARE
220 0.1 Analytical Balance, NTEP-Approved, 220 g x 0.1 mg
₹1,048,000.00INR / Each