Mettler-Toledo Excellence Melting Point Systems
Simple and automatic determination of melting points
  • One Click™ user interface with large color display allows you to start measurements with the click of a button
  • Reliable results with automatic detection of transmission intensity and digital image analysis
  • High resolution video captures unattended measurements for easy recall at any time
The Excellence melting point system is everything you need for the simple, automatic determination of melting points – run up to three samples at once, statistical analysis, and printable results. The temperature accuracy of +/- 0.2°C from ambient to 200°C and +/- 0.5°C from 200 to 300°C and compliance with standards such as Ph. Eur. and USP ensure accurate results. Models MP30 and MP55 detect the melting point up to 300°C and MP70 up to 350°C. The short heating and cooling times allow more analyses to be performed in a shorter period.
2,910,000.00 - ₹3,668,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Description
Availability Pricing
Mettler Toledo MP30 Excellence Melting Point System with 3 Melting Points 2302913 Mfr # MP30
COMPARE
3 min 300 Excellence Melting Point System with 3 Melting Points
₹2,910,000.00INR / Each
Mettler Toledo MP55 Excellence Melting Point System with 3 Melting Points with 2 Slip Points 2302914 Mfr # 30578101
COMPARE
3 min 300 Excellence Melting Point System with 3 Melting Points with 2 Slip Points
₹3,668,000.00INR / Each