Mettler Toledo EVA Coulometric Karl Fischer Titrators
Accurately and efficiently determine the water content for ultra-low (1 ppm to 5%) samples
  • Fast repeatable measurements without sacrificing accuracy
  • Electrochemically generated iodine helps measure extremely low water levels
  • Automatic solvent exchange of reduces contact with chemicals
  • Sealed, easy-to-clean hybrid cell streamlines cleaning, reducing the risk of contamination
  • Easy-to-understand One Click™ touchscreen user interface
  • Save shortcuts to minimize errors and save time
  • Flexible data export via USB ports or Ethernet
Ensure the quality of your product by accurately determining the water content in your sample. The EVA Karl Fischer titrators simplify sample handling and eliminate the risk of human error, delivering highly reproducible and accurate results within a matter of minutes. Coulometric titrators analyze liquids, soluble solids, and insoluble solids (EVA C3 only) with water content between 1 ppm to 5% with accuracy and efficiency.
A combination of the Fast Forecasting Amperometric (FFA™) control algorithm, a fast-response digital sensor, and state-of-the-art dispensing technology ensure fast and accurate results. Coulometric Karl Fischer titrators generate iodine electrochemically from iodide, allowing for precise quantification of low water levels. Use the large color touchscreen to operate the simple-to-use interface to customize shortcuts and simply daily tasks.
Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick, connect to a printer via the USB port, or network export via Ethernet.
4 PRODUCT OPTIONS
Compareitem specifications
Applications Special Features
Availability Pricing
Mettler Toledo 30988510 EVA C1 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm 3333222 Mfr # 30988510
COMPARE
Liquids and soluble solids Generator electrode without diaphragm
₹3,226,000.00INR / Each
Mettler Toledo 31028041 EVA C1 Coulometric Karl Fischer Titrator, Generator Electrode with Diaphragm 3333223 Mfr # 31028041
COMPARE
Liquids and soluble solids Generator electrode with diaphragm 
₹3,226,000.00INR / Each
Mettler Toledo 30988512 EVA C3 Coulometric Karl Fischer Titrator, Generator Electrode without Diaphragm 3333224 Mfr # 30988512
COMPARE
Liquids, soluble solids, and insoluble solids Generator electrode without diaphragm
₹4,300,000.00INR / Each
Mettler Toledo 31028043 EVA C3 Coulometric Karl Fischer Titrator, Generator Electrode with Diaphragm  3333225 Mfr # 31028043
COMPARE
Liquids, soluble solids, and insoluble solids Generator electrode with diaphragm 
₹4,300,000.00INR / Each