Memmert ICH Climate Chambers
Specially designed for testing of pharmaceuticals according to ICH
  • Climate friendly eco models use refrigerant carbon dioxide for cooling
  • No icing, drying out of samples, or dehumidification of the working chamber
  • Motor-driven forced air circulation ensures homogenous temperature distribution
ICH climate chambers are made for testing pharmaceuticals according to ICH, as well as testing the stability of cosmetics and food. Samples are protected through cooling aggregate and heating being situated outside the working chamber in the air jacket. The air jacket surrounds the entire chamber, ensuring precise quick and precise temperature control. The air circulation is also motor driven and adjustable in 10% steps, ensuring very homogenous temperature distribution. It can heat the interior up to 60 °C and cool it down to -10 °C (without humidity and without light) or +10 °C (with humidity). Humidification of the interior is achieved by evaporating water from a tank at a set rate by means of a hot-air generator on the rear side of the appliance. The sterile hot air is introduced into the interior above the fan and mixed with the air current. Humidity is reduced through condensation, using Peltier cooling modules on the rear side of the appliance. Ice that might build up due to dehumidification is automatically defrosted in cycles. Eco models use refrigerant carbon dioxide (R744), which is not flammable, toxic, or ozone depleting.
Each climate chamber is supplied with AtmoCONTROL software on a USB stick. With AtmoCONTROL software, programs can be transferred to the appliance and protocols can be read out.
4,053,000.00 - ₹9,930,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
Memmert ICH 110 Climate Chamber, 108 L, 115 VAC 5220056 Mfr # ICH 110 115V
COMPARE
115 Climate Chamber, 108 L, 115 VAC
₹4,053,000.00INR / Each
Memmert ICH 260 Climate Chamber, 256 L, 115 VAC 5220057 Mfr # ICH 260 115V
COMPARE
115 Climate Chamber, 256 L, 115 VAC
₹6,089,000.00INR / Each
Memmert ICH 750 Climate Chamber, 749 L, 115 VAC 5220058 Mfr # ICH 750 115V
COMPARE
115 Climate Chamber, 749 L, 115 VAC
₹8,282,000.00INR / Each
Memmert ICH 110 L Climate Chamber with Illumination, 108 L, 115 VAC 5220059 Mfr # ICH 110 L 115V
COMPARE
115 Climate Chamber with Illumination, 108 L, 115 VAC
₹5,117,000.00INR / Each
Memmert ICH 260 L Climate Chamber with Illumination, 256 L, 115 VAC 5220061 Mfr # ICH 260 L 115V
COMPARE
115 Climate Chamber with Illumination, 256 L, 115 VAC
₹7,703,000.00INR / Each
Memmert ICH 750 L Climate Chamber with Illumination, 749 L, 115 VAC 5220062 Mfr # ICH 750 L 115V
COMPARE
115 Climate Chamber with Illumination, 749 L, 115 VAC
₹9,930,000.00INR / Each