Memmert HPP eco Constant Climate Chambers
Cost-effective energy-efficient Peltier technology
  • No compressor, no coolants, maintenance free!
  • Perfectly suited for stability tests, storage in controlled environment and conditioning
  • Multiple over/under temperature protection features with audible and visual alarms
These quiet constant climate chambers are unbeatable in energy efficiency. Energy-saving Peltier heating/cooling system provides forced air ventilation and multiple elements can be controlled individually to ensure homogeneous temperature and humidity distribution inside the chamber.
The easy-to-use ControlCOCKPIT with twin high-definition TFT color displays provides control and display of all operating parameters including temperature, humidity, air flap, timer, fan, set point, and actual temperature. Versatile menu functions allow easy setting of language, time zones, daylight saving time, and choice between Celsius and Fahrenheit. Integrated USB port on control panel for easy uploading of programs, reading out protocol logs, and activating the User-ID function.
Chambers feature internal data logger with storage capacity for 10 years. Set point WAIT function guarantees process time does not start until temperature is reached at all measuring points. Ethernet interface for uploading and implementing programs and for online logging. AtmoCONTROL software supplied on USB stick for programming and documentation.
Stainless steel interior and exterior construction make cleanup easy. Three layer insulation, inner glass door, changeable hinges and recessed grips (lockable casters on 26.5, 37.4, 48, and 75.6 cu ft models) provide durability and user friendly options.
10,063,000.00 - ₹16,735,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC) Shelf Positions
Availability Pricing
Memmert HPP1060eco Constant Climate Chamber, 37.4 cu ft, 115 VAC 5220052 Mfr # HPP1060 ECO 115V
COMPARE
37.4 115 14
₹10,063,000.00INR / Each
Memmert HPP1400eco Constant Climate Chamber, 48 cu ft, 115 VAC 5220053 Mfr # HPP1400 ECO 115V
COMPARE
48 115 28
₹13,634,000.00INR / Each
Memmert HPP2200eco Constant Climate Chamber, 75.6 cu ft, 230 VAC 5220054 Mfr # HPP2200 ECO 230V
COMPARE
75.6 230 42
₹16,735,000.00INR / Each