Megger DLRO100 Series Digital Low-Resistance Ohmmeters Accessories

Test leads and other accessories to customize your ohmmeter
Compatible for use with the Megger® 100 A Digital Low-Resistance Ohmmeters 20026-25 through -30.
82,400.00 - ₹223,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Megger Lead Set for DLRO100 Ohmmeter, CAT IV 600 V, 5 m WW-20026-89 Mfr # 1004-448
COMPARE
Lead Set for DLRO100 Ohmmeter, CAT IV 600 V, 5 m
₹223,000.00INR / Each
Megger Kelvin Lead Set for DLRO100 Ohmmeter, CAT IV 600 V, 5 m WW-20026-93 Mfr # 1005-634
COMPARE
Kelvin Lead Set for DLRO100 Ohmmeter, CAT IV 600 V, 5 m
₹223,000.00INR / Each
Megger DC Clamp for DLRO100 Ohmmeter, MCPD 100L WW-20026-96 Mfr # 1005-622
COMPARE
DC Clamp for DLRO100 Ohmmeter, MCPD 100L
₹82,400.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat