Mark-10 R04 Series Miniature Tension/Compression Force Sensors

Ultra-compact size
  • Measures tensile and compressive forces
  • Weight: 0.015 lb (0.007 kg)
  • Warranty: 3 years
Extremely compact force measurement sensor for tensile and compressive loads in applications with limited space. The sensors feature outstanding mechanical overload protection and threaded holes on two sides for the mounting of hook attachments or other implements.
496,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Capacity (Newtons)
Availability Pricing
Mark-10 MR04-2 Force Sensor, 2 lbF / 32 ozF / 1 kgF / 1000 gF / 10 N 2530254 Mfr # MR04-2M
COMPARE
2 10
₹496,000.00INR / Each
Mark-10 MR04-5 Force Sensor, 5 lbF / 80 ozF / 2.5 kgF / 2500 gF / 25 N 2530255 Mfr # MR04-5
COMPARE
5 25
₹496,000.00INR / Each
Mark-10 MR04-20 Force Sensor, 20 lbF / 320 ozF / 10 kgF / 10000 gF / 100 N 2530257 Mfr # MR04-20
COMPARE
20 100
₹496,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat