Mark-10 Professional Digital Force Gauges

Advanced solution for demanding force measurement applications
  • High-speed 14,000 Hz sampling rate
  • Continuous data capture of up to 5,000 data points, at up to 14,000 Hz, downloadable to a PC
  • USB, RS-232, Mitutoyo, and analog outputs
  • Sample break detection – automatically zeroes, saves, outputs, and stops a Mark-10 test stand upon sample break
  • 1st / 2nd peak detection
  • Five units of measurement, plus COF and user-defined units
  • Programmable footswitch command string
  • Averaging, external trigger, real-time, and peak modes
  • Programmable set points, with audiovisual indicators and outputs
  • Password protection
Professional digital force gauges are designed with advanced features for your most demanding tension and compression measurement applications, up to 500 lb (2500 N). Gauges feature an industry-leading sampling rate of 14,000 Hz with ±0.1% accuracy, producing quick and reliable results for rapid testing applications.

These gauges offer all the functions of the M5-series Premium force gauges plus several additional features, including high-speed continuous data capture and storage, with memory for up to 5000 readings, at an acquisition rate of up to 14,000 Hz. Units also feature programmable footswitch sequencing, break detection, 1st / 2nd peak detection. They include coefficient of friction (COF) and user-defined units of measurement, with the ability to export data from the storage buffer to optional MEASUR™gauge software for advanced analysis and graphing.

For enhanced productivity, gauges also feature automatic data output, data storage, and zeroing functions upon the completion of break detection, averaging, external trigger, and 1st / 2nd peak detection. All gauges are compatible with Mark-10 test stands, including the ESM301, to enable functions such as break testing, tensile testing, compression testing, dynamic load holding, PC control capability, and more.
Call For Price

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Resolution (lbs)
Availability Pricing
Mark-10 M7-012 Force Gauge, 0.12 lbF / 2 ozF / 50 gF / 0.5 N / 500 mN 2530261 Mfr # M7-012
COMPARE
0.12 2E-05
Call For Price
Mark-10 M7-05 Force Gauge, 0.5 lbF / 8 ozF / 250 gF / 2.5 N / 2500 mN 2530263 Mfr # M7-05
COMPARE
0.5 0.0001
Call For Price
Mark-10 M7-2 Force Gauge, 2 lbF / 32 ozF / 1 kgF / 1000 gF / 10 N 2530264 Mfr # M7-2
COMPARE
2 0.0005
Call For Price
Mark-10 M7-5 Force Gauge, 5 lbF / 80 ozF / 2.5 kgF / 2500 gF / 25 N 2530265 Mfr # M7-5
COMPARE
5 0.001
Call For Price
Mark-10 M7-10 Force Gauge, 10 lbF / 160 ozF / 5 kgF / 5000 gF / 50 N 2530266 Mfr # M7-10
COMPARE
10 0.002
Call For Price
Mark-10 M7-20 Force Gauge, 20 lbF / 320 ozF / 10 kgF / 10000 gF / 100 N 2530267 Mfr # M7-20
COMPARE
20 0.005
Call For Price
Mark-10 M7-50 Force Gauge, 50 lbF / 800 ozF / 25 kgF / 25000 gF / 250 N 2530268 Mfr # M7-50
COMPARE
50 0.01
Call For Price
Mark-10 M7-200 Force Gauge, 200 lbF / 3200 ozF / 100 kgF / 1000 N / 1 kN 2530270 Mfr # M7-200
COMPARE
200 0.05
Call For Price
Mark-10 M7-500 Force Gauge, 500 lbF / 8000 ozF / 250 kgF / 2500 N / 2.5 kN 2530271 Mfr # M7-500
COMPARE
500 0.1
Call For Price
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat