Luxor Tub Shelf Utility Carts
Safety transport heavy loads in deep tub shelves
  • Four full-swivel 4” (300-400 lb) or 5" (500 lb) casters, two with locking brakes, make transporting goods easy
  • HDPE shelves and PVC legs never rust, dent, scratch, or stain
The tub shelf utility carts constructed of durable HDPE, feature deep tub walls to safely transport heavy loads and keep contents contained. The ergonomic push handle molded into the top shelf and the smooth rolling, heavy-duty casters, allow you to easily maneuver the cart over any flooring.
34,800.00 - ₹63,900.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Height (in) Number of Shelves
Availability Pricing
Luxor Tub Shelf Utility Cart, Large, with Two 2-3/4" Tub Shelves, Black 4760030 Mfr # TC11-B
COMPARE
400 37.5 2
₹53,600.00INR / Each
Luxor Tub Shelf Utility Cart, Large, with Two 2-3/4" Tub Shelves, Gray 4760032 Mfr # TC11-G
COMPARE
400 37.5 2
₹63,900.00INR / Each
Luxor Tub Shelf Utility Cart with Two 2-3/4" Tub Shelves, Black 4760034 Mfr # STC11-B
COMPARE
300 37.5 2
₹36,500.00INR / Each
Luxor Tub Shelf Utility Cart with Two 2-3/4" Tub Shelves, Gray 4760036 Mfr # STC11-G
COMPARE
300 37.5 2
₹34,800.00INR / Each
Luxor Tub Shelf Utility Cart with Three 2-3/4" Tub Shelves, Black 4760038 Mfr # STC111-B
COMPARE
300 39.25 3
₹36,300.00INR / Each
Luxor EC11-B Tub Shelf Utility Cart with Two 2-1/2" Tub Shelves, Black 4760039 Mfr # EC11-B
COMPARE
400 34.25 2
₹59,900.00INR / Each
Luxor Tub Shelf Utility Cart with Three 2-3/4" Tub Shelves, Gray 4760040 Mfr # STC111-G
COMPARE
300 39.25 3
₹50,300.00INR / Each
Luxor Tub Shelf Utility Cart, Heavy Duty, with Two 2-1/2" Tub Shelves, Black 4760098 Mfr # XLC11-B
COMPARE
500 33 2
₹47,800.00INR / Each
Showing 1- 8 of 8