Luxor Modular Room Divider Wall System
Easily divide larger areas into quiet functional spaces
  • Dampen sound—PET dividers are rated to absorb 85% of ambient noise
  • Panels are constructed of a mix of PET and frosted acrylic to absorb noise without blocking natural light
  • Choose between starter and add-on walls to create a custom configuration to accommodate any workspace
The Luxor modular divider wall system is designed to divide larger areas into quieter, more functional spaces. The system features a mix of sound-absorbent PET and semi-transparent frosted acrylic inner panels that you can arrange to optimize privacy, reduce noise, and enhance natural light. The PET panels absorb 85% of ambient noise (NRC 0.85), are tackable for posting notes or pictures, and can be cleaned with up to 75% isopropyl alcohol. The frosted acrylic panels enhance privacy without blocking natural light, can double as a whiteboard when used with water-soluble dry-erase markers, and can be cleaned with mild soap and water.
Starter walls are freestanding and can be used as a stand-alone solution or connected to an add-on wall to create a custom configuration. Each wall features a built-in cable management tray to help keep cords organized, and its durable aluminum frame with sturdy steel feet to ensure stability and years of trouble-free use.
95,000.00 - ₹144,000.00INR / Pkg of 2
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dimension Height (in) Width (in)
Availability Pricing
Luxor Modular Room Divider Wall System, Starter Wall, 53" x 48"; 2/Pk 4760080 Mfr # MW-5348-FCG
COMPARE
48" H Starter Wall 48 53
₹97,100.00INR / Pkg of 2
Luxor Modular Room Divider Wall System, Add-On Wall, 53" x 48" 4760081 Mfr # MW-5348-XFCG
COMPARE
48" H Add-On Wall 48 53
₹95,000.00INR / Each
Luxor Modular Room Divider Wall System, Starter Wall, 53" x 70"; 2/Pk 4760082 Mfr # MW-5370-FCG
COMPARE
70" H Starter Wall 70 53
₹114,000.00INR / Pkg of 2
Luxor Modular Room Divider Wall System, Add-On Wall, 53" x 70"; 2/Pk 4760083 Mfr # MW-5370-XFCG
COMPARE
70" H Add-On Wall 70 53
₹119,000.00INR / Pkg of 2
Luxor Modular Room Divider Wall System, Starter Wall, 70" x 48"; 2/Pk 4760084 Mfr # MW-7048-FCG
COMPARE
48" H Starter Wall 48 70
₹123,000.00INR / Pkg of 2
Luxor Modular Room Divider Wall System, Add-On Wall, 70" x 48"; 2/Pk 4760085 Mfr # MW-7048-XFCG
COMPARE
48" H Add-On Wall 48 70
₹111,000.00INR / Pkg of 2
Luxor Modular Room Divider Wall System, Starter Wall, 70" x 70" 4760086 Mfr # MW-7070-FCG
COMPARE
70" H Starter Wall 70 70
₹144,000.00INR / Each
Luxor Modular Room Divider Wall System, Add-On Wall, 70" x 70"; 2/Pk 4760087 Mfr # MW-7070-XFCG
COMPARE
70" H Add-On Wall 70 70
₹137,000.00INR / Pkg of 2
Showing 1- 8 of 8