Lovibond® TB350 Portable Turbidity Meters
Laboratory accuracy in a portable meter with the light-absorbing trap (BLAC®), which eliminates stray light and provides extremely accurate results down to 0.01 NTU
  • Reads up to 4000 NTU without the need for dilutions
  • Fast Settling Mode increases accuracy of readings with large or heavy particles
  • Superior data management - easily save and recall measurement data sets
  • Touchscreen interface for ease of operation
The Lovibond® TB Series of turbidimeters offers simplistic operation combined with intelligent instrument engineering to provide an unparalleled level of accuracy in turbidity measurement.
Ideal for field and environmental testing, the TB350 turbidimeter delivers the most reliable measurements for low range to high range samples without sacrificing accuracy. Featuring the patent-pending Multipath 90° BLAC® sensor technology, the optical system is engineered with dual detectors to deliver a ratio reading which mitigates common measurement stability issues. The detection angle responds most consistently to a wide range of particle sizes and shapes in any given sample. The new BLAC technology stands for: Backscattered Light Absorbing Cavity. The light-absorbing trap eliminates stray light and provides extremely accurate results for low turbidity down to 0.01 NTU.
The intuitive touchscreen interface makes it easy to perform procedures and interpret results. With function applications, you can navigate between settings and perform them efficiently. This user interface helps eliminate common frustrations and prevents errors.
The data logging capabilities allow you to record the testing location, operator’s identification, time, and date along with the measurement. Stored data can be transferred to a computer via USB.
552,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Min Turbidity Description
Availability Pricing
Lovibond TB350 WL White Light Turbidimeter Kit 9959985 Mfr # 194310
COMPARE
4000 NTU (FNU) 0.01 NTU (FNU) White Light Turbidimeter Kit
₹552,000.00INR / Each
Lovibond TB350 IR Infrared Light Turbidimeter Kit 9959986 Mfr # 194300
COMPARE
4000 NTU (FNU) 0.01 NTU (FNU) Infrared Light Turbidimeter Kit
₹552,000.00INR / Each