Labnet BioPette Plus Autoclavable, Adjustable Volume Pipettes, Single-Channel
Autoclavable, comfortable and accurate
  • Superior accuracy and reproducibility
  • Convenient thumbwheel and push-button volume selection
  • Universal shaft accepts most pipette tips
This line of pipettes is ergonomically designed for comfort and engineered to be lightweight and well balanced—use in either your right or left hand. Large digital display provides enhanced visibility. Chemical-resistant, maintenance-free seals and finely polished pistons provide unsurpassed accuracy sample after sample. Adjustable digital micrometer is slightly recessed to prevent unintentional changes and is protected from the effects of thermal expansion by the insulated pipettor body. The tip ejector is ergonomically positioned to reduce fatigue during prolonged use, is adjustable to accommodate various styles of tips, and is removable for pipetting into narrow tubes. Tip ejector’s stainless steel construction resists corrosion, rust, and breakage. autoclavable.
56,900.00 - ₹63,400.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
Labnet BioPette P3940-2 Autoclavable Pipette, adjustable vol, 0.1-2.0 uL WW-24601-00 Mfr # P3942-2
COMPARE
Yes 0.002 0.0001
₹60,900.00INR / Each
Labnet BioPette P3940-10 Autoclavable Pipette, adjustable vol, 0.5 - 10.0 uL WW-24601-02 Mfr # P3942-10
COMPARE
Yes 0.01 0.0005
₹60,900.00INR / Each
Labnet BioPette P3940-20 Autoclavable Pipette, adjustable vol, 2-20 uL WW-24601-04 Mfr # P3942-20
COMPARE
Yes 0.02 0.002
₹56,900.00INR / Each
Labnet BioPette P3940-100 Autoclavable Pipette, adjustable vol, 10-100 uL WW-24601-06 Mfr # P3942-100
COMPARE
Yes 0.1 0.01
₹57,100.00INR / Each
Labnet BioPette P3940-200 Autoclavable Pipette, adjustable vol, 20-200 uL WW-24601-08 Mfr # P3942-200
COMPARE
Yes 0.2 0.02
₹56,900.00INR / Each
Labnet BioPette P3940-1000 Autoclavable Pipette, adjustable vol, 100-1000 uL WW-24601-10 Mfr # P3942-1000
COMPARE
Yes 1 0.1
₹57,100.00INR / Each
Labnet BioPette P3940-5000 Autoclavable Pipette, adjustable vol, 1000 - 5000 uL WW-24601-12 Mfr # P3942-5000
COMPARE
Yes 5 1
₹62,600.00INR / Each
Labnet BioPette P3940-10000 Autoclavable Pipette, adjustable vol, 1000 - 10000 uL WW-24601-14 Mfr # P3942-10000
COMPARE
Yes 10 1
₹63,400.00INR / Each
Showing 1- 8 of 8