Labconco Zero-Pressure Weather Caps
Designed for quick installation
  • Constructed of durable PVC
The Zero Pressure Weathercap is made of strong, corrosion-resistant PVC and is designed to connect directly to Labconco Thermoplastic Duct. The cap adds little static pressure to the exhaust system and allows for vertical discharge of effluent air for dispersion away from the building. It is not compatible with metal ductwork.
274,000.00 - ₹318,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Ductwork Diameter (in) Exterior Height (in) Description
Availability Pricing
Labconco Zero Pressure Weather Cap, PVC, 10" Diameter 0907579 Mfr # 7095100
COMPARE
10 48 Zero Pressure Weather Cap, PVC, 10" Diameter
₹274,000.00INR / Each
Labconco Zero Pressure Weather Cap, PVC, 12" Diameter 0907679 Mfr # 5622100
COMPARE
12 56 Zero Pressure Weather Cap, PVC, 12" Diameter
₹318,000.00INR / Each