Labconco XPert TXE Equipment Enclosure
Designed to address the needs of pharmaceutical research and quality control laboratories
Equipment enclosure provides a safe, ventilated space for keeping powder-generating apparatus contained. Its tall 42'' interior height accommodates equipment such as pill presses, grinders, crushers and sieves. HEPA-filtered air is returned to the laboratory. Rear baffle with zones of perforations promotes horizontal laminar airflow. The baffle sections pivot down or may be removed for cleaning. The interior height of 42” accommodates tall equipment such as pill presses, grinders, crushers and sieves. Enclosure has a built-in fan with speed control and vibration-isolation supports.
Enclosure also features a blower with 40% reserve capacity to overcome filter loading, and a HEPA exhaust filter which is 99.99% efficient. The HEPA filter traps particles 0.3 micron in size so that clean air returns to the laboratory. The HEPA filter is a true bag-in/bag-out filter disposal system. Enclosure is static-dissipative and features a durable epoxy-coated metal exterior, stainless steel interior sides and integral work surface, and safety glass doors. A pre-wired fluorescent lamp features a separate switche for blower and light. Adjustable-height doors are made of 1/4'' (0.6 cm) thick tempered safety glass. Door hinges are reversible to accommodate right or left side placement. Magnets keep doors securely closed.
Models with Guardian Airflow Monitor include a monitor that continuously monitors airflow with a green LED that glows when airflow is within set point range and a red LED that glows and an audible alarm that sounds when airflow is outside set point range.
Models with Guardian 1000 Digital Airflow Monitor include a monitor that continuously monitors airflow and provides a digital readout of actual face velocity. An audible/ visual alarm alerts the operator when airflow is outside set point range. RS232 communications port allows connection to user-supplied computer.
All models conform to the following regulations and standards:
SEFA 1-2005
Modified ASHRAE 110-95
ANSI Z9.5-2003
UL 3101-1/61010-1
CAN/CSA C22.2 No. 1010.1
CE Conformity Marking (230 volt models
All models require a supporting base. Order base below from accessores
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interior Height (in) Interior Width (in) Description
Availability Pricing
Labconco Xpert TXE 3892000 Equipment Enclosure, 115 Volt 3351200 Mfr # 3892000
COMPARE
42 22.5 Equipment Enclosure, 115 V
Call For Price
Labconco Xpert TXE 3892001 Equipment Enclosure with Guardian Airflow Monitor, 115 VOLT 3351202 Mfr # 3892001
COMPARE
42 22.5 Equipment Enclosure with Guardian Airflow Monitor, 115 V
Call For Price
Labconco Xpert TXE 3892002 with Guardian1000 Digital Airflow Monitor,115Volts 3351204 Mfr # 3892002
COMPARE
42 22.5 Equipment Enclosure with Guardian 1000 Digital Airflow Monitor, 115 V
Call For Price
Labconco Xpert TXE 3892020 Equipment Enclosure, 230 VOLT 3351206 Mfr # 3892020
COMPARE
42 22.5 Equipment Enclosure, 230 V
Call For Price
Labconco Xpert TXE 3892021 Equipment Enclosure with Guardian Airflow Monitor, 230 VOLT 3351208 Mfr # 3892021
COMPARE
42 22.5 Equipment Enclosure with Guardian Airflow Monitor, 230 V
Call For Price
Labconco Xpert TXE 3892022 with Guardian1000 Digital Airflow Monitor,230Volts 3351210 Mfr # 3892022
COMPARE
42 22.5 Equipment Enclosure with Guardian 1000 Digital Airflow Monitor, 230 V