This product is now discontinued, View Related Products
Labconco XPert Bulk Powder Filtered Systems
Protect yourself during powder transfer operations
  • Stainless steel work surface with front spill trough to catch spills
  • Push-button hydraulic lift for source container
XPert® bulk powder filter systems protect you from hazardous airborne particulates and fumes generated during powder transfer operations. Systems feature built-in blowers with dynamically balanced, vibration-isolated impellers with speed control, pre-wired fluorescent lamp with switch, and exhaust HEPA filter efficient on particled 0.3 microns. Models also available with center and right-side source container access openings. Call our Application Specialists for information regarding enclosures and filtered stations.
What’s included: transfer caddy with 3"-dia casters, 2"-square tubular steel base with leveling feet, exhaust HEPA filter, built-in blower with speed control, pre-wired fluorescent lamp and switches for blower and light, digital airflow
6,500.00 - ₹162,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Exterior Width (in) Description
Availability Pricing
Labconco Xpert 3961500 5' Bulk Powder Filtered System WW-33509-00 Mfr # 3961500
COMPARE
60 5' Bulk Powder Filtered System
Call For Price
Labconco Xpert 3961520 5' Bulk Powder Filtered System 230V Left side access WW-33509-05 Mfr # 3961520
COMPARE
60 5' Bulk Powder Filtered System 230V Left side access
Call For Price
Labconco Xpert 3961600 6' Bulk Powder Filtered System 115V Left side access WW-33509-10 Mfr # 3961600
COMPARE
72 6' Bulk Powder Filtered System 115V Left side access
Call For Price
Labconco Xpert 3961620 6' Bulk Powder Filtered System 230V Left side access WW-33509-15 Mfr # 3961620
COMPARE
72 6' Bulk Powder Filtered System 230V Left side access
Call For Price
Labconco 3981300 19.62" Retaining Ring, Type 304 Stainless Steel WW-33509-70 Mfr # 3981300
COMPARE
19.62\" Retaining Ring, Type 304 Stainless Steel
₹145,000.00INR / Each
Labconco 3981301 17.5" Retaining Ring, Type 304 Stainless Steel WW-33509-72 Mfr # 3981301
COMPARE
17.5\" Retaining Ring, Type 304 Stainless Steel
₹162,000.00INR / Each
Labconco 3981302 14.5" Retaining Ring, Type 304 Stainless Steel WW-33509-74 Mfr # 3981302
COMPARE
14.5\" Retaining Ring, Type 304 Stainless Steel
₹162,000.00INR / Each
Labconco 3981303 11.5" Retaining Ring, Type 304 Stainless Steel WW-33509-76 Mfr # 3981303
COMPARE
11.5\" Retaining Ring, Type 304 Stainless Steel
₹162,000.00INR / Each
Labconco 3981100 Safety Sleeves Package of 100 WW-33509-80 Mfr # 3981100
COMPARE
Safety Sleeves Package of 100
₹29,200.00INR / Each
Labconco 3981101 Safety Sleeves Package of 500 WW-33509-82 Mfr # 3981101
COMPARE
Safety Sleeves Package of 500
₹121,000.00INR / Each
Labconco 3981200 Retaining Bands package of 5 WW-33509-84 Mfr # 3981200
COMPARE
Retaining Bands package of 5
₹6,500.00INR / Each
Showing 1- 11 of 11