Labconco WaterPro BT Water Purification Systems
Compact benchtop design produces both Type I and Type III (RO) water!
  • Set up in minutes—simply connect to tap supply, insert the filter pack, and power on
  • Integrated 6-liter storage tank holds Type III RO-purified water
  • Quiet—50 dB—and compact enough to be shared between labs, set up on a benchtop, or wall mounted
WaterPro BT systems deliver up to 10 L/day of ASTM Type I water direct from a tap feed water supply. Ideal for lab applications such as trace elemental analysis, mixing or diluting acid/base solutions, and media preparation.
In addition, ASTM Type III RO-purified water is produced at a rate of 3 L/hr* and stored in an integrated 6-L tank. Ready and waiting for glassware rinsing, reagent preparation, and other noncritical laboratory water needs.
Compact, lightweight, and simple to install, these systems can be easily transported and shared among labs. All water filtration technologies-activated carbon filtration, deionization, and RO-are combined into one convenient twin-cartridge pack that snaps into place (order required filter pack 99290-48 separately).
User-friendly color LCD shows water quality, system status, and maintenance or alarm messages. Plus, it can be tilted for easy viewing from any angle, even if system is wall mounted.
Keypad allows quick access to display settings, water quality unit of measure, resistivity set point, and temperature values. It also provides a means to set the Type I water volume to be dispensed, dispensing time, flow rate calibration, and tank level calibration.
For further purification, choose from two optional final filters that attach to the dispensing valve. The hollow fiber filter removes all particles and bacteria larger than 0.22 micron. The ultrafilter cartridge removes pyrogens (pathogens, RNase, and DNase).
*Feedwater conductivity must be 100 µS or better.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Power (VAC) With UV Lamp
Availability Pricing
Labconco 9015020 WaterPro BT Water Purification System; 115V 9929040 Mfr # 9015020
COMPARE
WaterPro BT Water Purification System; 115V 115 No
Call For Price
Labconco 9015220 WaterPro BT Water Purification System, Remote Dispense; 115V 9929042 Mfr # 9015220
COMPARE
WaterPro BT Water Purification System, Remote Dispense; 115V 115 No
Call For Price
Labconco 9015120 WaterPro BT Water Purification System, UV Lamp; 115V 9929044 Mfr # 9015120
COMPARE
WaterPro BT Water Purification System, UV Lamp; 115V 115 Yes
Call For Price
Labconco 9015340 WaterPro BT Water Purification System, Remote Dispense, UV Lamp; 240V 9929047 Mfr # 9015340
COMPARE
WaterPro BT Water Purification System, Remote Dispense, UV Lamp; 240V 240 Yes