Labconco Replacement Fast-Freeze Flasks, Amber Glass
Borosilicate glass construction withstands extreme temperatures and high vacuum
  • Amber-colored glass protects light-sensitive samples
  • Flat bottom and wide mouth design provides easy loading and cleaning
Use with snap-on tops and flask adapters to attach flasks to freeze dryer valves.
47,700.00 - ₹76,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Labconco FreeZone Fast-Freeze Flask, Amber, Glass, 300 mL 0335328 Mfr # 7542601
COMPARE
300
₹47,700.00INR / Each
Labconco FreeZone Fast-Freeze Flask, Amber, Glass, 600 mL 0335329 Mfr # 7542801
COMPARE
600
₹67,000.00INR / Each
Labconco FreeZone Fast-Freeze Flask, Amber, Glass, 750 mL 0335330 Mfr # 7542701
COMPARE
750
₹69,900.00INR / Each
Labconco FreeZone Fast-Freeze Flask, Amber, Glass, 900 mL 0335331 Mfr # 7542901
COMPARE
900
₹76,500.00INR / Each