Labconco RapidVap Vertex Dry Evaporator
Quick evaporation of up to 50 samples at the same time
  • Built-in nitrogen regulator with independent on/off control with five rows of 10 nozzles — conserves energy during partial runs
  • Touch-screen LCD allows up to 10 user-set programs that may be stored in memory and provides display during the run
  • Compact benchtop design with small footprint
Unlike water bath heaters, the dry block heater requires less maintenance, no distilled water or additives, and add no potential source of contamination. Ideal for preparation of samples in a variety of applications including drug discovery, environmental testing, agrochemistry, and forensic evidence processing. Quality construction and reliable performance are backed by a one-year warranty.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Plug Type Power (VAC)
Availability Pricing
Labconco 7320020 RapidVap Vertex Evaporator, NEMA 5-15P Plug; 115 VAC, 60 Hz 1304099 Mfr # 7320020
COMPARE
NEMA 5-15P 115
Call For Price
Labconco 7320030 RapidVap Vertex Evaporator, Shuko Plug; 230 VAC, 50/60 Hz 2685313 Mfr # 7320030
COMPARE
Schuko 230