Labconco Purifier Vertical Clean Benches
Remove particles from room air to supply Class 100 air to your bench working area
Vertical clean benches are typically used for plant tissue culture, media plate preparation, and electronics inspection. Order optional epoxy work surface for added chemical resistance and spill containment, or telescoping base stands for ergonomic enclosure height adjustment.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Width (in) Power (VAC)
Availability Pricing
Labconco 3970220 Vertical Clean Bench, 2-ft, 230V 0909640 Mfr # 3970220
COMPARE
24 230
Call For Price
Labconco 3970320 Vertical Clean Bench, 3-ft, 230V 0909655 Mfr # 3970320
COMPARE
36 230