Labconco® Logic® Vue Class II Biosafety Enclosures
Spacious Class II enclosures provide ample room for a variety of instruments
  • Meets NSF/ANSI 49 requirements for personnel and product protection
  • Continuous airflow monitoring for user safety
  • Two fully opening sashes provide easy equipment loading
  • Ergonomically designed with built-in knee space
  • Spacious working areas: 42.2"H x 31"D (107.2 x 78.7 cm)
  • Durable 304 stainless steel (SS) interior construction
  • Password protection prevents accidental setting changes
The Logic® Vue Class II biosafety enclosures are built with your safety and comfort in mind. A HEPA supply and exhaust filters provide >99.99% efficiency (at 0.3 µm) for ISO Class 5/Grade A air or better. Constant Airflow Profile (CAP) system continuously monitors and adjusts airflow to contain biohazards within the enclosure ensuring user safety.
View status information and configure settings using the touchpad controller with a full-color LCD screen. The controller uses MyLogic™ operating system that communicates important status information such as filter life, blower status, alerts, alarms, safety interlock*, and data connectivity. Remotely monitor the status using the USB connection or dry contact relays.
The spacious enclosures accommodate a variety of automated instruments. Multiple pass-throughs allow safe passages of cables, cords, and service fixtures. Durable polycarbonate side windows and fully opening front and rear sashes offer easy accessibility and visibility from all sides. Get unrestricted access to full working area via an 8" (20.3 cm) front lower sash opening. Work surface height is adjustable from 31.5 to 37.5" (80 and 95.3 cm) in 1" increments (2.5 cm); a convenient knee well offers comfort while working seated. A 133 footcandle (1432 lux) LED lighting illuminates work surface for maximum visibility.
Chemically resistant epoxy powder-coated steel exterior and 304 SS interior construction offer excellent durability. A SpillStopper work surface tray accommodates instruments up to 250 lb (113.4 kg) per every 2 feet (61 cm). International packaging models require transportation by high cube shipping containers. Customize your biosafety enclosures with a variety of accessories—order separately.
7,737,000.00 - ₹9,677,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interior Width (in) Power (VAC) Description
Availability Pricing
Labconco Logic® Vue Class II Enclosure, 6-ft Wide; 115V, 30A WW-33517-80 Mfr # 3300600
COMPARE
73.8 115 6-ft Wide Class II Enclosure; 115V, 30A
₹9,677,000.00INR / Each
Labconco Logic® Vue Class II Enclosure, 6-ft Wide; 230V, 20A WW-33517-81 Mfr # 3300601
COMPARE
73.8 230 6-ft Wide Class II Enclosure; 230V, 20A
₹9,677,000.00INR / Each
Labconco Logic® Vue Class II Enclosure, 4-ft Wide; 115V, 30A WW-33517-87 Mfr # 3300400
COMPARE
49.8 115 4-ft Wide Class II Enclosure; 115V, 30A
₹7,737,000.00INR / Each
Labconco Logic® Vue Class II Enclosure, 4-ft Wide; 230V, 20A WW-33517-88 Mfr # 3300401
COMPARE
49.8 230 4-ft Wide Class II Enclosure; 230V, 20A
₹7,737,000.00INR / Each
Labconco Logic® Vue Class II Enclosure with International Packaging, 4-ft Wide; 115V, 30A WW-33517-89 Mfr # 3300410
COMPARE
49.8 115 4-ft Wide Class II Enclosure with International Packaging; 115V, 30A
₹7,737,000.00INR / Each
Labconco Logic® Vue Class II Enclosure with International Packaging, 4-ft Wide; 230V, 20A WW-33517-90 Mfr # 3300411
COMPARE
49.8 230 4-ft Wide Class II Enclosure with International Packaging; 230V, 20A
₹7,737,000.00INR / Each
Labconco Logic® Vue Class II Enclosure with International Packaging, 6-ft Wide; 115V, 30A WW-33517-91 Mfr # 3300610
COMPARE
73.8 115 6-ft Wide Class II Enclosure with International Packaging; 115V, 30A
₹9,677,000.00INR / Each
Labconco Logic® Vue Class II Enclosure with International Packaging, 6-ft Wide; 230V, 20A WW-33517-92 Mfr # 3300611
COMPARE
73.8 230 6-ft Wide Class II Enclosure with International Packaging; 230V, 20A
₹9,677,000.00INR / Each
Showing 1- 8 of 8