Lab-Grade Glass Dewar Flasks
Highly efficient laboratory-grade Dewars feature low heat conductivity and high vacuum. Annealed borosilicate glass reduces risk of flask implosion. Weighted base reduces spillage. Resilient plastic mesh covering cushions the flask and ensures a firm grip. Vented polyethylene stopper (included) minimizes evaporation.
35,700.00 - ₹116,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Volume (mL)
Availability Pricing
COMPARE
350
₹35,700.00INR / Each
COMPARE
665
₹37,900.00INR / Each
COMPARE
1000
₹40,200.00INR / Each
COMPARE
1900
₹101,000.00INR / Each
COMPARE
4300
₹116,000.00INR / Each