Konica Minolta Illuminance Meters
Illuminance meters provide highly accurate and reliable measurements
  • Conform to Class AA JIS C 1609-1:2006 and DIN 5032 Part 7 Class B standards
  • Provides multi-point measurement capabilities – 2 to 30 points
  • Accurately measures PWM-controlled light sources
  • Model T-10MA includes a min receptor for small hard to reach places
  • Easy-to-read LCD display
These illuminance meters provide measurement functions that include illuninance (lx), illuninance difference (lx), illuminance ratio (%), integrated illuminance (lx-h), integration time (h), and average illuminance (lx). Meters feature a silicon photocell receptor that provides highly accurate measurements and includes a USB port for easily downloading measurement data. Use the optional software, part number 50537-09, to import your measurements into Excel® for report generation and analysis. Software sold separately.
589,000.00 - ₹765,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Measurement (Lux) Sensor Description
Availability Pricing
Konica Minolta T-10A Illuminance Light Meter 5053707 Mfr # A58U-213 (T-10A)
COMPARE
299900 Built-in Illuminance Light Meter
₹589,000.00INR / Each
Konica Minolta T-10MA Illuminance Meter with 1M Receptor 5053708 Mfr # A58U-223 (T-10MA)
COMPARE
299900 Remote Illuminance Meter with 1M Receptor
₹765,000.00INR / Each