Koflo Standard PVC Schedule 80 Pipe In-Line Static Mixers
Mix fluids as you pump them through a pipe line
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
27,800.00 - ₹73,500.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 1/2-80-4-6-2 Standard PVC schedule 80 pipe mixer; 1/2" NPT(M), 6 elements 0466900 Mfr # 1/2-80-4-6-2
COMPARE
5 0.5" NPT(M) 6
₹27,800.00INR / Each
Koflo 3/4-80-4-6-2 Standard PVC schedule 80 pipe mixer; 3/4" NPT(M), 6 elements 0466902 Mfr # 3/4-80-4-6-2
COMPARE
7 0.75" NPT(M) 6
₹28,500.00INR / Each
Koflo 1-80-4-6-2 Standard PVC schedule 80 pipe mixer; 1" NPT(M), 6 elements 0466904 Mfr # 1-80-4-6-2
COMPARE
10 1" NPT(M) 6
₹31,600.00INR / Each
Koflo 3/4-80-4-12-2 Standard PVC schedule 80 pipe mixer; 3/4" NPT(M), 12 elements 0466908 Mfr # 3/4-80-4-12-2
COMPARE
14 0.75" NPT(M) 12
₹47,700.00INR / Each
Koflo 1-80-4-12-2 Standard PVC schedule 80 pipe mixer; 1" NPT(M), 12 elements 0466910 Mfr # 1-80-4-12-2
COMPARE
19 1" NPT(M) 12
₹51,800.00INR / Each
Koflo 1.5-80-4-6-2 Standard PVC schedule 80 pipe mixer, 1-1/2" NPT(M), 6 elements 0466978 Mfr # 1.5-80-4-6-2
COMPARE
15 1.5" NPT(M) 6
₹38,000.00INR / Each
Koflo 1.5-80-4-12-2 Standard PVC schedule 80 pipe mixer, 1-1/2" NPT(M), 12 elements 0466979 Mfr # 1.5-80-4-12-2
COMPARE
26 1.5" NPT(M) 12
₹63,100.00INR / Each
Koflo 2-80-4-12-2 Standard PVC schedule 80 pipe mixer, 2" NPT(M), 12 elements 0466981 Mfr # 2-80-4-12-2
COMPARE
33 2" NPT(M) 12
₹73,500.00INR / Each
Koflo 2-80-4-6-2 Standard PVC schedule 80 pipe mixer, 2" NPT(M), 6 elements 0466983 Mfr # 2-80-4-6-2
COMPARE
18 2" NPT(M) 6
₹44,300.00INR / Each
Showing 1- 9 of 9