Koflo PVC Mixers with Injection Port
Mix fluids as you pump them through a pipe line
  • Economical, long-lasting, and more convenient than most motorized mixers
  • In-line mixers prevent overmixing and undermixing, and resist fouling in your pipes or tubes
  • Virtually maintenance-free and need no spare parts
In-line mixers are more convenient than most motorized mixers. Instead of mixing fluids with a motorized mixer before pumping, you can mix fluids while you pump them through a pipe line. In-line mixers prevent overmixing and undermixing, and resist fouling in your pipes or tubes. They are virtually maintenance free and need no spare parts.
In-line mixers provide efficient mixing and can withstand large pressure changes — up to 1500 psi with smallest sizes. Mixing elements in pipe and tube mixers are securely attached to the mixer walls, so internal pressure changes cannot collapse elements and stop flow.
These PVC in-line mixers have an injection port designed for blending light-viscosity chemicals into a water stream. Each mixer is constructed of schedule 80 PVC and has four fixed mixing elements. Connections are 150-lb raised face flanged ends.
Note: These mixers are designed for high flow rates. They use four elements instead of six to minimize pressure loss.
61,100.00 - ₹259,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 1-80-4-4-9I PVC Mixer with Injection Port, 1" Line size 0466860 Mfr # 1-80-4-4-9.I
COMPARE
12 0.5" NPT(M) 4
₹61,100.00INR / Each
Koflo 1.5-80-4-4-9I PVC Mixer with Injection Port, 1-1/2" Line size 0466862 Mfr # 1.5-80-4-4-9.I
COMPARE
16 0.5" NPT(M) 4
₹66,000.00INR / Each
Koflo 2-80-4-4-9I PVC Mixer with Injection Port, 2" Line size 0466864 Mfr # 2-80-4-4-9I
COMPARE
19 0.75" NPT(M) 4
₹76,500.00INR / Each
Koflo 2.5-80-4-4-9I PVC Mixer with Injection Port, 2-1/2" Line size 0466866 Mfr # 2.5-80-4-4-9I
COMPARE
22 0.75" NPT(M) 4
₹120,000.00INR / Each
Koflo 3-80-4-4-9I PVC Mixer with Injection Port, 3" Line size 0466868 Mfr # 3-80-4-4-9I
COMPARE
27 0.75" NPT(M) 4
₹138,000.00INR / Each
Koflo 4-80-4-4-9I PVC Mixer with Injection Port, 4" Line size 0466870 Mfr # 4-80-4-4-9I
COMPARE
32 0.75" NPT(M) 4
₹183,000.00INR / Each
Koflo 6-80-4-4-9I PVC Mixer with Injection Port, 6" Line size 0466872 Mfr # 6-80-4-4-9I
COMPARE
41 1" NPT(M) 4
₹259,000.00INR / Each
Showing 1- 7 of 7