Koflo 316 Stainless Steel In-Line Pipe Mixers
Mix fluids as you pump them through a pipe line
  • Offers 100% mixed output
  • Industrial grade materials ensure long service life
  • Durability under demanding conditions
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
31,300.00 - ₹148,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 1/4-40-3-6-2 Pipe Mixer, 316 Stainless Steel, 1/4 inch NPT(M), 6 elements 0466905 Mfr # 1/4-40-3-6-2
COMPARE
3 0.25" NPT(M) 6
₹31,300.00INR / Each
Koflo 1/4-40-3-12-2 Pipe Mixer, 316 Stainless Steel, 1/4 inch NPT(M), 12 elements 0466907 Mfr # 1/4-40-3-12-2
COMPARE
6 0.25" NPT(M) 12
₹47,700.00INR / Each
Koflo 3/8-40-3-6-2 Pipe Mixer, 316 Stainless Steel, 3/8 inch NPT(M), 6 elements 0466909 Mfr # 3/8-40-3-6-2
COMPARE
4 0.375" NPT(M) 6
₹35,500.00INR / Each
Koflo 3/8-40-3-12-2 Pipe Mixer, 316 Stainless Steel, 3/8 inch NPT(M), 12 elements 0466911 Mfr # 3/8-40-3-12-2
COMPARE
8 0.375" NPT(M) 12
₹50,500.00INR / Each
Koflo 1/2-40-3-6-2 Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 6 elements 0466912 Mfr # 1/2-40-3-6-2
COMPARE
5 0.5" NPT(M) 6
₹38,100.00INR / Each
Koflo 3/4-40-3-6-2 Pipe Mixer, 316 stainless steel, 3/4" NPT(M), 6 elements 0466914 Mfr # 3/4-40-3-6-2
COMPARE
7 0.75" NPT(M) 6
₹40,200.00INR / Each
Koflo 1-40-3-6-2 Pipe Mixer, 316 stainless steel, 1" NPT(M), 6 elements 0466916 Mfr # 1-40-3-6-2
COMPARE
9 1" NPT(M) 6
₹43,100.00INR / Each
Koflo 1/2-40-3-12-2 Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 12 elements 0466918 Mfr # 1/2-40-3-12-2
COMPARE
10 0.5" NPT(M) 12
₹55,200.00INR / Each
Koflo 3/4-40-3-12-2 Pipe Mixer, 316 stainless steel, 3/4" NPT(M), 12 elements 0466920 Mfr # 3/4-40-3-12-2
COMPARE
13 0.75" NPT(M) 12
₹58,800.00INR / Each
Koflo 1-40-3-12-2 Pipe Mixer, 316 stainless steel, 1" NPT(M), 12 elements 0466922 Mfr # 1-40-3-12-2
COMPARE
17 1" NPT(M) 12
₹67,000.00INR / Each
Koflo 1.5-40-3-6-2 Pipe Mixer, 316 stainless steel, 1-1/2" NPT(M), 6 elements 0466923 Mfr # 1.5-40-3-6-2
COMPARE
13 1.5" NPT(M) 6
₹58,200.00INR / Each
Koflo 1.5-40-3-12-2 Pipe Mixer, 316 stainless steel, 1-1/2" NPT(M), 12 elements 0466924 Mfr # 1.5-40-3-12-2
COMPARE
26 1.5" NPT(M) 12
₹90,900.00INR / Each
Koflo 2-40-3-6-2 Pipe Mixer, 316 stainless steel, 2" NPT(M), 6 elements 0466925 Mfr # 2-40-3-6-2
COMPARE
17 2" NPT(M) 6
₹90,900.00INR / Each
Koflo 2-40-3-12-2 Pipe Mixer, 316 stainless steel, 2" NPT(M), 12 elements 0466926 Mfr # 2-40-3-12-2
COMPARE
33 2" NPT(M) 12
₹148,000.00INR / Each
Showing 1- 14 of 14