Koehler Penetrometer Cones for Manual and Digital Penetrometers
Compatible with both manual and digital penetrometers
  • Precision machined cones and needles for ASTM and related methods
Use with penetrometers to determine the consistency of petroleum products.
193,000.00 - ₹381,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material
Availability Pricing
Koehler K20800 Penetrometer Precision Cone, Magnesium with Stainless Steel Tip, 102.5 g 5986088 Mfr # K20800
COMPARE
Magnesium with Stainless Steel Tip
₹298,000.00INR / Each
Koehler K20000 Penetrometer Precision Cone, Brass with Stainless Steel Tip, 102.5 g 5986090 Mfr # K20000
COMPARE
Brass with Stainless Steel Tip
₹193,000.00INR / Each
Koehler K20900 Penetrometer Precision Cone, Aluminum, Quarter Scale, 2.48 g 5986096 Mfr # K20900
COMPARE
Aluminum
₹381,000.00INR / Each