Kinetic Vibration-Free Platforms
Enhance the performance of your balances with a vibration-free tabletop
  • Low profile passive- or active-air designs
  • Excellent horizontal isolation
  • ISO Class 5 (formerly Class 100) cleanroom compatible
These platforms reduce vibration and shock when using precision testing equipment, such as balances. There are two configurations, the passive-air and active-air platforms. The passive-air design requires an optional hand pump if an air source is not available. It requires periodic manual leveling. The active-air design requires an air source (not included) for use. It is a self-leveling unit and recommended for regular use. The load capacity on all models is 200 lb.
594,000.00 - ₹780,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kinetic active-air Vibration-Free Platform, White Plastic Laminate 20" x 24" Active-Air 0101945 Mfr # 2212-01-11
COMPARE
Vibration-Free Platform, White Plastic Laminate 20" x 24" Active-Air
₹780,000.00INR / Each
Kinetic passive-air Vibration-Free Platform, Black Plastic Laminate 16" x 19" Passive Air 0101949 Mfr # 2210-04-13
COMPARE
Vibration-Free Platform, Black Plastic Laminate 16" x 24" Passive Air
₹594,000.00INR / Each
Kinetic Passive-Air Vibration-Free Platform, White Plastic Laminate; 20" x 24" 0101951 Mfr # 2210-01-11
COMPARE
Vibration-Free Platform, White Plastic Laminate 20" x 24" Passive Air
₹615,000.00INR / Each