Cole-Parmer® Snap Caps
No need for tool with quick and easy snap caps
  • Snap caps are ideal for use when a crimping tool is not available
  • Constructed of durable polypropylene or polyethylene
  • Select from a variety of caps and septa materials that fit your application and needs
Snap Top Caps can be snapped on and off by hand which eliminates the need for metal crimping and decapping tools. Snap caps are available in polypropylene (PP) or polyethylene (PE) with various septum materials. Select septa with slit to prevent a vacuum from forming inside the vials.
Caps are available in different colors, septum materials, and open or closed configurations to suit your application.
4,900.00 - ₹128,000.00INR / Pkg of 1000
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap color Cap Diameter (mm) Septum Type
Availability Pricing
COMPARE
Transparent 11 PTFE red/silicone white/PTFE red
₹34,300.00INR / Pkg of 1000
COMPARE
Transparent 11 Silicone White/PTFE red
₹27,400.00INR / Pkg of 1000
COMPARE
Transparent 11 Silicone white/PTFE blue with star burst slit
₹36,400.00INR / Pkg of 1000
COMPARE
Black 11 Silicone White/PTFE red
₹31,400.00INR / Pkg of 1000
COMPARE
Light Blue 11 PTFE
₹24,900.00INR / Pkg of 1000
COMPARE
Transparent 11 Natural rubber/TEF
₹26,400.00INR / Pkg of 1000
COMPARE
Transparent 11 Rubber/PTFE
₹32,000.00INR / Pkg of 1000
COMPARE
Blue 11 Natural rubber/TEF
₹27,600.00INR / Pkg of 1000
COMPARE
Blue 11 Natural rubber/TEF
₹31,400.00INR / Pkg of 1000
COMPARE
Blue 11 Red rubber/PTFE beige
₹33,100.00INR / Pkg of 1000
COMPARE
Red 11 Natural rubber/TEF
₹31,400.00INR / Pkg of 1000
COMPARE
Blue 11 PTFE red/silicone white/PTFE red
₹53,000.00INR / Pkg of 1000
COMPARE
Red 11 PTFE red/silicone white/PTFE red
₹53,000.00INR / Pkg of 1000
COMPARE
Transparent 11 UltraClean silicone white/PTFE red
₹40,700.00INR / Pkg of 1000
COMPARE
Blue 11 UltraClean silicone white/PTFE red
₹45,500.00INR / Pkg of 1000
COMPARE
Blue 11 UltraClean silicone white/PTFE blue
₹45,500.00INR / Pkg of 1000
COMPARE
Red 11 UltraClean silicone white/PTFE red
₹45,500.00INR / Pkg of 1000
COMPARE
Transparent 11 UltraClean pre-slit silicone white/PTFE blue
₹39,900.00INR / Pkg of 1000
COMPARE
White 11 Silicone White/PTFE red
₹40,700.00INR / Pkg of 1000
COMPARE
Red 11 Silicone white/PTFE blue with pre-slit
₹39,000.00INR / Pkg of 1000
Showing 1 - 20 of 30