Katanax® FluxPenser® Flux Dispenser
Gain efficiency, precision, and excellent reproducibility in sample and flux weighing
  • Adds flux to the desired weight within 1 mg accuracy
  • Automates weighing task, avoiding repetitive strain
  • Dispense granular or micro bead flux for optimal flexibility
Automatically calculate and accurately dispense the required amount of flux to the sample with a flux dosing dispenser. Two operating modes make it easy to accommodate your dispensing needs. Select absolute mode for high-accuracy flux dosing and weighing; relative mode lets you obtain a desired flux/sample ratio by weighing your sample then adding flux to the desired ratio. Silo is provided to facilitate flux reloading, while minimizing contamination and moisture exposure. Weight data can be transferred to most LIMS systems over standard communication protocols.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Power (VAC)
Availability Pricing
Environmental Express Katanax® FluxPenser® Flux Dispenser 0457889 Mfr # XFP-01
COMPARE
1 mg 110 / 220
Call For Price
COMPARE
115 / 230