Kapton Type HN film
Kapton® Type HN film is a tough, aromatic polyimide film, exhibiting an excellent balance of physical, chemical, and electrical properties over a wide temperature range, particularly at unusually high temperatures. It has been used successfully in applications at temperatures as low as -452°F (-269°C) and as high as 752°F (400°C). HN film can be laminated, metallized, punched, formed or adhesive coated.
3,800.00 - ₹13,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Thickness (mil)
Availability Pricing
COMPARE
12 Polymide 1
₹4,000.00INR / Each
COMPARE
12 Polymide 2
₹3,800.00INR / Each
COMPARE
12 Polymide 5
₹13,000.00INR / Each